Learn More
Total Base Number Standard, ASTM D 2896 (nominal value: 3.01 mg KOH/g), 1 x 50g, Paragon Scientific™
Specifically manufactured for the verification of analytical instruments used to determine base number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D2896 / IP 276.
Brand:  Paragon Scientific TBN3
Product Code. 16000450
Description
Description
  • Base Number (TBN) tested and certified in accordance with ASTM D2896 / IP 276
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Supplied as single volume packs or as triple packs
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
  • Manufactured in the United Kingdom
  • 24 month shelf life
Specifications
Specifications
1 x 50 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Total Base Number Standard, ASTM D 2896 (nominal value: 3.01 mg KOH/g), 1 x 50g, Paragon Scientific™ > Quantity: 1 x 50g
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Nominal values quoted. Certified Values will vary from batch to batch.