Learn More
Thermo Scientific™ Oxoid™ Ampicillin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0003B
329.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10671655
  • 554.00 NOK / Pack of 250
Explore more special offers
Description
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Ampicillin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ampicillin
10µg
AMP
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with CLSI, DIN, BSAC, EUCAST, SFM guidelines
Oxoid Ampicillin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6248246 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberCT0003B
Lot Number6244938 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog NumberCT0003B
Lot Number6243654 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberCT0003B
Lot Number6230099 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog NumberCT0003B
Lot Number6227134 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberCT0003B
Product Title
Thermo Scientific™ Oxoid™ Ampicillin Antimicrobial Susceptibility discs > 10μg