Learn More
Thermo Scientific™ Oxoid™ Gentamicin Antimicrobial Susceptibility Discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand: Thermo Scientific™ CT0024B
314.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Gentamycin Antimicrobial Susceptibility discs.
- Easy-to-read labeling
- Shatterproof, spring-loaded cartridges
- End-of-cartridge indicator
- Reproducible performance
Ordering Alerts:
50 Discs/Cartridge; 5 Cartridges/Package. Sold by 5 Cartridges/Package only.
Specifications
Gentamicin | |
10µg | |
CN | |
Antimicrobial Susceptibility | |
50 Discs/Cartridge |
Complies with CLSI, DIN, BSAC, EUCAST guidelines | |
Oxoid Gentamicin Antimicrobial Susceptibility Discs | |
Antimicrobial Susceptibility Discs | |
5 Cartridges/Pk. | |
50 Discs/Cartridge |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6251129 | Certificate of Analysis | 25/04/2025 | CT0024B |
6245029 | Certificate of Analysis | 25/04/2025 | CT0024B |
6217811 | Certificate of Analysis | 27/03/2025 | CT0024B |
6230101 | Certificate of Analysis | 27/03/2025 | CT0024B |
6241909 | Certificate of Analysis | 21/03/2025 | CT0024B |