Learn More
Memmert™ Factory Calibration (3 Temperatures) for UN30
Calibration at Memmert factory for three temperature value.
Brand: Memmert™ UN30D00128
This item is not returnable.
View return policy
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
UN30 Oven | |
Measuring point chamber center | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Memmert™ Factory Calibration (3 Temperatures) for UN30 > For Use With: UN30