Learn More
Sartorius™ Silicone Gaskets
233.00 NOK
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
10613772
|
Sartorius™
6980570 |
20.5 x 26.5 x 0.5 mm |
233.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10679602
|
Sartorius™
6980569 |
14.9 mm |
233.00 NOK
Pack of 10 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||||
Description
Specifications
Silicone | |
Gasket | |
10 Pack |