Learn More
Sartorius™ Silicone Gaskets
Brand:  Sartorius™ 6980570
171.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10613772
  • 233.00 NOK / Pack of 10
In Stock
to see stock.
Explore more special offers
Specifications
10 Pack
20.5 x 26.5 x 0.5 mm
Silicone
Gasket
Product Title
Sartorius™ Silicone Gaskets > Gasket; Dimensions: 20.5 x 26.5 x 0.5mm