Learn More
Sartorius™ Silicone Gaskets
Brand:  Sartorius™ 6980569
171.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10679602
  • 233.00 NOK / Pack of 10
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Specifications
10 Pack
14.9 mm
Silicone
Gasket
Documents
One moment while we fetch your results.
Product Title
Sartorius™ Silicone Gaskets > Gasket; Dimensions: 14.9mm