Learn More
Kartell™ Polypropylene Pipette Tips
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
277.00 NOK - 1360.00 NOK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | CE / IVD |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Packaging | Color | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Packaging | Color | Material | Price | Quantity & Availability | ||
12533287
|
Kartell™
0097100 |
2 to 300 μL | Bulk | Neutral | Polypropylene |
277.00 NOK
Pack of 1000 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||
12503267
|
Kartell™
0974600 |
0.5 to 20 μL | Refill | Neutral | Polypropylene |
935.00 NOK
Pack of 4800 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||
10358271
|
Kartell™
0964600 |
0.1 to 10 mL | Rack | Natural | Polypropylene |
1360.00 NOK
Pack of 960 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||
10469371
|
Kartell™
0964800 |
1000 to 5000 μL | Rack | Light Blue | Polypropylene |
1345.00 NOK
Pack of 200 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
||
Description
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
- Disposable tips are also available racked
Specifications
Autoclavable | |
CE / IVD | |
Polypropylene |