Learn More
Kartell™ Polypropylene Pipette Tips
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
277.00 NOK - 1360.00 NOK
Specifications
Autoclavable Autoclavable
Certifications/Compliance CE / IVD
Material Polypropylene
View More Specs
Products 4
Product Code Brand Volume (Metric) Packaging Color Material Price Quantity & Availability  
Product Code Brand Volume (Metric) Packaging Color Material Price Quantity & Availability  
12533287
View Documents Promotion Details
Kartell™
0097100
2 to 300 μL Bulk Neutral Polypropylene
277.00 NOK
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
12503267
View Documents Promotion Details
Kartell™
0974600
0.5 to 20 μL Refill Neutral Polypropylene
935.00 NOK
Pack of 4800
View alternative products
Save Up to 
»
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
10358271
View Documents Promotion Details
Kartell™
0964600
0.1 to 10 mL Rack Natural Polypropylene
1360.00 NOK
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 04-10-2024
to see stock.
Add to basket
10469371
View Documents Promotion Details
Kartell™
0964800
1000 to 5000 μL Rack Light Blue Polypropylene
1345.00 NOK
Pack of 200
View alternative products
Save Up to 
»
Estimated Shipment: 10-10-2024
Description
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable tips are also available racked
Specifications
Specifications
Autoclavable
CE / IVD