Learn More
Kartell™ Polypropylene Pipette Tips
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand: Kartell™ 0964600
This item is not returnable.
View return policy
Description
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
- Disposable tips are also available racked
Specifications
0.1 to 10 mL | |
Autoclavable | |
Rack | |
Gilson™ |
Natural | |
CE / IVD | |
Rack | |
Polypropylene |