Learn More
Kartell™ Polypropylene Pipette Tips
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand:  Kartell™ 0964600
 View more versions of this product
Product Code. 10358271
  • 1360.00 NOK / Pack of 960
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable tips are also available racked
Specifications
Specifications
0.1 to 10 mL
Autoclavable
Rack
Gilson™
Natural
CE / IVD
Rack
Polypropylene