missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Oleoyl Coenzyme A Dipotassium Salt
Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.
583.00 NOK - 3420.00 NOK
Specifications
Color | White |
---|---|
Content And Storage | -20°C |
Form | Powder |
Molecular Weight (g/mol) | 1031.986 |
Product Type | Biochemicals |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394099
|
MP Biomedicals™
0210090401 |
1 mg |
583.00 NOK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304109
|
MP Biomedicals™
0210090405 |
5 mg |
1727.80 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314109
|
MP Biomedicals™
0210090410 |
10 mg |
3420.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Coenzyme A functions as an acyl group carrier, acetyl-CoA
- Coenzyme A functions as an acyl group carrier, acetyl-CoA
Specifications
White | |
Powder | |
Biochemicals |
-20°C | |
1031.986 | |
≥90% |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.