Learn More
MP Biomedicals™ Oleoyl Coenzyme A Dipotassium Salt
Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.
Brand:  MP Biomedicals™ 0210090410
 View more versions of this product
Product Code. 11314109
  • 3420.00 NOK / 10mg
Explore more special offers
Description
Description
  • Coenzyme A functions as an acyl group carrier, acetyl-CoA
  • Coenzyme A functions as an acyl group carrier, acetyl-CoA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Powder
1031.986
≥90%
-20°C
10 mg
Biochemicals
Special Offers
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.