Learn More
MP Biomedicals™ Oleoyl Coenzyme A Dipotassium Salt
Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.
Brand: MP Biomedicals™ 0210090405
This item is not returnable.
View return policy
Description
- Coenzyme A functions as an acyl group carrier, acetyl-CoA
- Coenzyme A functions as an acyl group carrier, acetyl-CoA
Specifications
White | |
Powder | |
1031.986 | |
≥90% |
-20°C | |
5 mg | |
Biochemicals |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.