missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Necrosulfonamide
Necroptosis inhibitor
1720.00 NOK - 6365.00 NOK
Specifications
Purity | >98% |
---|---|
Product Type | Necrosulfonamide |
Molecular Weight (g/mol) | 461.47 |
Inhibitors | Necrosulfonamide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755688
|
Tocris Bioscience™
5025/50 |
50 mg |
6365.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700138
|
Tocris Bioscience™
5025/10 |
10 mg |
1720.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Necroptosis inhibitor. Blocks mixed lineage kinase domain-like protein (MLKL), a critical substrate of receptor-interacting serine-threonine kinase 3 (RIP3) during necrosis. Prevents MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors.Specifications
>98% | |
Necrosulfonamide | |
461.47 | |
Necrosulfonamide |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ Necrosulfonamide