Learn More
Tocris Bioscience™ Necrosulfonamide
Necroptosis inhibitor
Brand: Tocris Bioscience™ 5025/50
4882.81 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Necrosulfonamide is a necroptosis inhibitor. Blocks mixed lineage kinase domain-like protein (MLKL), a critical substrate of receptor-interacting serine-threonine kinase 3 (RIP3) during necrosis. Prevents MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors. Binds and inhibits gasdermin D. Inhibits pyroptosis.Specifications
>98% | |
Necrosulfonamide | |
Necrosulfonamide |
50 mg | |
461.47 |
For Research Use Only