Learn More
Tocris Bioscience™ Necrosulfonamide
Necroptosis inhibitor
Brand:  Tocris Bioscience™ 5025/10
 View more versions of this product
Product Code. 18700138
  • 1720.00 NOK / 10mg
Explore more special offers
Description
Description
Necrosulfonamide is a necroptosis inhibitor. Blocks mixed lineage kinase domain-like protein (MLKL), a critical substrate of receptor-interacting serine-threonine kinase 3 (RIP3) during necrosis. Prevents MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors. Binds and inhibits gasdermin D. Inhibits pyroptosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>98%
Necrosulfonamide
Necrosulfonamide
10 mg
461.47
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tocris Bioscience™ Necrosulfonamide > 10mg