missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse SLAM/CD150 ELISA Kit

Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant.

5130.00 NOK - 40770.00 NOK

Specifications

Accession Number 4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1
Assay Range 0.06 to 12 ng/mL
Assay Sensitivity 0.06 ng/mL
Conjugate Biotin
Product Type ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17185669
View Documents
Invitrogen™
EM70RBX5
5 x 96 Tests
26530.00 NOK
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
17164923
View Documents
Invitrogen™
EM70RB
96 Tests
5130.00 NOK
Pack of 96
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
 
18557964
View Documents
Invitrogen™
EM70RBX10
10 x 96 Tests
40770.00 NOK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse SLAM/CD150.

CD150 (SLAM, Signaling Lymphocyte Activation Molecule) belongs to the immunoglobulin gene superfamily, and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells. Moreover, CD150 is thought to interact with SH2D1A and with PTPN11 via its cytoplasmic domain. Mutations CD150 gene may be associated with X-linked lympho-proliferative disease (XLP).
Specifications

Specifications

4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1
0.06 ng/mL
ELISA
Q9QUM4
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Research Use Only
CD150
4 hr 45 min
0.06 to 12 ng/mL
Biotin
Colorimetric Microplate Reader
Slamf1
<10%
HRP
2°C to 8°C
1 hr 20 min
Videos
SDS
Documents

Documents

Product Certifications