Learn More
Invitrogen™ Mouse SLAM/CD150 ELISA Kit
Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant.
Brand:  Invitrogen™ EM70RB
 View more versions of this product
Product Code. 17164923
  • 5130.00 NOK / Pack of 96
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Description
Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse SLAM/CD150.
CD150 (SLAM, Signaling Lymphocyte Activation Molecule) belongs to the immunoglobulin gene superfamily, and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells. Moreover, CD150 is thought to interact with SH2D1A and with PTPN11 via its cytoplasmic domain. Mutations CD150 gene may be associated with X-linked lympho-proliferative disease (XLP).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1
0.06 ng/mL
ELISA
Colorimetric Microplate Reader
Slamf1
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.06 to 12 ng/mL
Biotin
Plasma, Serum, Supernatant
Q9QUM4
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
CD150
4 hr 45 min