missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse SLAM/CD150 ELISA Kit
Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant.
Brand: Invitrogen™ EM70RBX10
This item is not returnable.
View return policy
Description
Mouse SLAM/CD150 quantitates mouse SLAM/CD150 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse SLAM/CD150.
CD150 (SLAM, Signaling Lymphocyte Activation Molecule) belongs to the immunoglobulin gene superfamily, and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells. Moreover, CD150 is thought to interact with SH2D1A and with PTPN11 via its cytoplasmic domain. Mutations CD150 gene may be associated with X-linked lympho-proliferative disease (XLP).Specifications
4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; SLAMF1 | |
0.06 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Slamf1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.06 to 12 ng/mL | |
Biotin | |
Serum | |
Q9QUM4 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 µL; Plasma, 50 µL; Supernatant, 50 µL | |
CD150 | |
4 hr 45 min |