Learn More
STEMCELL Technologies Mouse Recombinant IL-10
Interleukin 10
2907.89 NOK - 8486.61 NOK
Specifications
Name B-TCGF, CSIF, Cytokine synthesis inhibitory factor, Interleukin-10, TGIF
Cell Type Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells; T Cells, CD4+; T Cells, Regulatory
Purity or Quality Grade ≥95%
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17109521
Encompass_Preferred
View Documents
STEMCELL Technologies
78079.1
50 μg
8486.61 NOK
50µg
View alternative products
Save Up to 
»
17199511
Encompass_Preferred
View Documents
STEMCELL Technologies
78079
10 μg
2907.89 NOK
10µg
View alternative products
Save Up to 
»
Description
Description
  • Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
  • All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
  • Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
  • It is produced by dendritic cells, macrophages, and CD4+ T regulatory cells, as well as mast cells, NK cells, neutrophils, and regulatory B cells, under specific stimulating conditions
  • IL-10 can inhibit the activation of certain immune cells while it promotes the function of B cells, and facilitates the healing process
  • Specifically, this cytokine is important for the function of T regulatory cells as it is a potent suppressor of effector T cell proliferation and cytokine production
  • Also, IL-10 produced by a subset of macrophages inhibits activation and production of pro-inflammatory cytokines by neighboring macrophages, thus allowing a level of self-regulation
Specifications
Specifications
B-TCGF, CSIF, Cytokine synthesis inhibitory factor, Interleukin-10, TGIF
Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells; T Cells, CD4+; T Cells, Regulatory
≥95%
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.