Learn More
STEMCELL Technologies Mouse Recombinant IL-10
Interleukin 10
Brand: STEMCELL Technologies 78079
This item is not returnable.
View return policy
Description
- Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
- All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
- Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
- It is produced by dendritic cells, macrophages, and CD4+ T regulatory cells, as well as mast cells, NK cells, neutrophils, and regulatory B cells, under specific stimulating conditions
- IL-10 can inhibit the activation of certain immune cells while it promotes the function of B cells, and facilitates the healing process
- Specifically, this cytokine is important for the function of T regulatory cells as it is a potent suppressor of effector T cell proliferation and cytokine production
- Also, IL-10 produced by a subset of macrophages inhibits activation and production of pro-inflammatory cytokines by neighboring macrophages, thus allowing a level of self-regulation
Specifications
B-TCGF, CSIF, Cytokine synthesis inhibitory factor, Interleukin-10, TGIF | |
10 μg | |
Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells; T Cells, CD4+; T Cells, Regulatory | |
≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.