Learn More
Motic™ Moticam A5
An ideal tool for those who need to stay flexible, the Moticam A5 has a high resolution above full HD while maintaining steady 30 fps. Take crisp pictures for documentation and public presentations.
Brand: Motic™ 1100600101302
Description
- C-mount for lens
- MotiConnect software is available for free download (with free updates included)
- Live Display Modes: 2592 x 1944 px, 2048 x 1536 px, 1920 x 1080 px
Specifications
Camera | |
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap | |
1/2.8 in. |
Moticam A | |
sCMOS | |
30 fps |