Learn More
Hirschmann™ Counting Chambers
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann Instruments™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8100103
 View more versions of this product
Product Code. 10200872
  • 759.00 NOK / Each
Only null left
Estimated Shipment: 28-07-2025
Description
  • Double net ruling
  • Available bright-lined or no, with or without clamps
  • Includes 2 cover glasses for haemocytometer
  • CE acc. IVD 98/79 EC
Specifications
Specifications
Counting Chambers, Neubauer
1Each
0.1mm
2 Haemocytometer Glass Covers
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Counting Chambers > Depth: 0.1mm; Ruled: Double Net