Learn More
Invitrogen™ EVOS™ M3000 Imaging System Product Code.: 30178687
Invitrogen™ EVOS™ M3000 Imaging System
Product Code. 30178687
EVOS M3000 Imaging System, Each
Click to view available options
Model:
EVOS M3000 Imaging System
Unit Size:
Each
Product Code. 30178687
Brand: Invitrogen™ AMF3000
This package includes the EVOS M3000 Imaging System (Cat. No. AMF3000) and a Rapid Exchange (REX) extended warranty.
The EVOS M3000 Imaging System is a fully integrated digital inverted benchtop microscope for two-color fluorescence, transmitted light, phase contrast, and color imaging. It is excellent for routine cell/tissue culture imaging applications. This system combines an easy user interface with powerful machine learning capabilities that allow users to quickly verify cell appearance, quality, and growth in seconds.
The EVOS M3000 offers you these important advantages:
  • Easy installation—no maintenance, assembly, alignment, or calibration
  • Automated real-time confluency analysis—includes pre-trained machine learning algorithm that can report cell confluency in the field of view at any time. As vessel is moved, confluency value automatically updates (in under a second on average).
  • User-defined two-color fluorescence—with addition of EVOS light cubes, two fluorescence channels can be acquired
  • Space-saving design—at 12 x 19 x 13 in. (W x D x H), system fits easily on lab bench or in cell culture hood
  • Network-capable—can be connected to local wireless network for seamless data transfer to OneDrive™ or Thermo Fisher Connect accounts. Alternatively, system offers on-board storage or export to USB.
  • Flexibility—choose from wide selection of EVOS-compatible objectives (1.25–60X) to best fit application and image quality needs
Real-time confluency analysis
Pre-trained using multiple common cell lines, the confluency tool automatically reports the confluency of the culture as it is moved over the stage in real-time. This removes inconsistency across multiple users and helps save time and energy compared to manual estimation.
Fully integrated imaging system
The EVOS M3000 Imaging System has a high-quality 10.3-inch LCD touchscreen display. The onboard computer and SSD (128 GB) are capable of processing and storing hours-worth of images in a single session or across multiple sessions. It contains a highly sensitive color CMOS camera (2,064 x 1,536-pixel resolution, 3.2 megapixels) for capturing high-quality 8-bit monochrome or color images in TIFF, PNG, and JPG formats, in addition to video capture in MP4 format. Adjust the contrast and saturation of images post-acquisition to produce stunning publication-ready images.
Versatile and highly configurable
With a four-position objective turret and the ability to acquire images in two fluorescence channels, transmitted light, phase contrast, and color, the system can be easily configured to help meet your needs. Choose from our broad range of high-quality long-working-distance and coverslip-corrected objectives for use with plastic vessels or glass sides. This system supports 1.25X to 60X magnification with achromat, fluorite, or apochromat correction, together with over a dozen LED light cubes. Image your cells the way you want to see them.
Easy to use and reliable
The EVOS M3000 Imaging System offers the simplicity of plug-and-play. It requires no warm-up or cool-down periods, allowing you to capture images quickly. The touchscreen display works with nitrile gloves, but the system also supports mouse and keyboard via USB connection. The EVOS M3000 is cleanroom compatible, capable of undergoing standard disinfection protocols including UV and hydrogen peroxide. Place the unit directly in a cell culture hood for rapid analysis of culture status and quality. An optional mechanical stage (Cat. No. AMEP5011) can be installed on the fixed stage for easier manipulation of vessels.
Our EVOS imaging systems are built from the ground up to help maximize performance and optimize workflow. You will be astonished at how easy this system is to operate and amazed at how extraordinary your images look on-screen. This system pushes the boundaries of what a manual imaging system can do.
Note: Objectives and light cubes are not included. They are sold separately.
Rapid Exchange Extended Warranty
Add warranty coverage to your existing manufacturer's limited warranty with the Rapid Exchange (REX) extended warranty. If your instrument needs repair, just call our Support Center, and we'll send you a factory-certified, refurbished, replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to the replacement unit.
The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.
Certain exclusions apply. Contact your Service and Support Representative for details at service.sales@thermofisher.com.
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Imaging System
Product Line EVOS
Camera CMOS
For Use With (Application) Imaging
Model EVOS M3000 Imaging System
Light Source LED
Magnification 1.25 to 60X
Objectives 4 Position Turret
Resolution 1920 x 1200 pixels
Stage Fixed (mechanical optional)
Display 10.1 in. LCD Touchscreen
Dimensions 11.7 x 12.7 x 19.1 in.
Weight (English) 18.4 lb.
Weight (Metric) 8.3 kg
Content And Storage
EVOS M3000 Imaging System
• 10.1-inch LCD touchscreen monitor, 1920 x 1080 pixel resolution
• Embedded PC and SSD with 8 GB RAM, gross 120 GB memory
• EPI Axis Shipping Restraint Block
• Cube Access Tool
• Light Cube Blank
EVOS M3000 Accessory Kit
• USB Wi-Fi Adapter (for wireless network connection to Connect applications)
• USB 3.0 Flash Drive (for image storage, preloaded with user documentation)
• Condenser Light Shield
• UV Light Shield Kit
• Polycarbonate Stage Plate
• Universal Power Supply, 12V, 5A
• Power Cord, Type B (North America)
• Block Condenser Slider (AMEP4688)
• Condenser Slider for 4X Objectives (AMEP4738)
• Diffusion Condenser Slider for Brightfield Applications (AMEPDFS1)
• EVOS Dust Cover
• Technical Support Card
* Note that one (1) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug:
AMEP4646 EVOS Power Cord, Type C (Europe)
AMEP4708 EVOS Power Cord, Type D (India/South Africa)
AMEP4645 EVOS Power Cord, Type G (United Kingdom)
AMEP4649 EVOS Power Cord, Type H (Israel)
AMEP4647 EVOS Power Cord, Type I (Australia)
AMEP4782 Power Cord, Type I (China, CCC certified)
AMEP4786 Power Cord Type J (Switzerland)
AMEP4648 EVOS Power Cord, Type L (Italy)
Quantity 1 system
Show More Show Less
For Research Use Only.
Product Content Correction
Product Title
Invitrogen™ EVOS™ M3000 Imaging System >