missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ John Guest™ Push-to-Connect Male Threaded Stem Adapters

Quick-disconnect convenience for a wide variety of tubing material including soft metals

Brand:  Cole-Parmer™ 34006-68

 View more versions of this product

Product Code. 11705828

  • 231.00 NOK / Pack of 10
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Meet NSF and FDA compliance for food-grade applications.
  • A single fitting offers secure connections and quick disconnection.
  • No need to purchase two fitting halves.
  • Simply push tubing past the O-ring to the internal tube stop.
  • Steel teeth on the collet grip tubing for leakproof connections.
  • To disconnect, press collet against fitting body to disengage steel teeth, tubing slides right out.
  • Fittings work with semi rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
  • To connect flexible tubing such as Tygon or silicone, order soft tube supports.
Specifications

Specifications

Push-to-Connect
Acetal
Air: -25°C, Water: 0°C
Autoclave or Ethylene Oxide
30 to 762 mm Hg
Air: 75°C, Water: 60°C
Autoclavable
Air: -14°F, Water: 33°F
3/8 in. x 1/4 in.
150 psi for air, 250 psi for water
Air: 167°F, Water: 140°F
Push-to-connect male threaded stem adapter, 3/8 in. x 1/4 in. NPTM, 10/pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ John Guest™ Push-to-Connect Male Threaded Stem Adapters > Push-to-connect male threaded stem adapter, 3/8 in

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.