missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JIB 04
Pan Jumonji inhibitor; active in vivo
2520.00 NOK - 9800.00 NOK
Specifications
Purity | >99% |
---|---|
Product Type | JIB 04 |
Molecular Weight (g/mol) | 308.76 |
Inhibitors | JIB 04 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756667
|
Tocris Bioscience™
4972/10 |
10 mg |
2520.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18768347
|
Tocris Bioscience™
4972/50 |
50 mg |
9800.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pan Jumonji histone demethylase inhibitor (IC50 values are 230, 340, 435, 445, 855 and 1100 nM for JARID1A, JMJD2E, JMJD2B, JMJD2A, JMJD3 and JMJD2C respectively). Exhibits negligible activity at PHD2 and TET1. Selectively blocks cancer cell growth in vitro and diminishes tumor growth in H358 and A549 mouse xenograft models in vivo. Prolongs survival in a mouse model of breast cancer.Specifications
>99% | |
JIB 04 | |
308.76 | |
JIB 04 |
For Research Use Only