Learn More
Lenz Laborglasintrumente Chromatographic Columns with Frit and Beaded Rim
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
819.00 NOK - 1130.00 NOK
Specifications
Material Duran Glass
View More Specs
Products 3
Product Code Brand Capacity (Metric) Reservoir Price Quantity & Availability  
Product Code Brand Capacity (Metric) Reservoir Price Quantity & Availability  
10683812
View Documents
Lenz Laborglasintrumente
05445706
430 mL
1035.00 NOK
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
11381612
View Documents
Lenz Laborglasintrumente
05450800
8 mL
1130.00 NOK
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
10413130
View Documents
Lenz Laborglasintrumente
05445702
35 mL
819.00 NOK
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • Chromatographic columns with sintered frit (porosity: 0)
  • With PTFE stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente Chromatographic Columns with Frit and Beaded Rim