Learn More
Bohlender™ BOLA™ Replacement Screw Caps, PPS
Replacement screw caps for sealing stirrer shafts.
133.00 NOK - 245.00 NOK
Specifications
For Use With (Application) Sealing Stirrer Shafts
Material PPS
View More Specs
Products 6
Product Code Brand Diameter (Metric) Shaft Diameter (English) Shaft Thread Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Shaft Diameter (English) Shaft Thread Size Price Quantity & Availability  
17157114
View Documents
Bohlender™
C425-82
6 mm 0.24 in. GL 18
171.00 NOK
Pack of 3
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
17167114
View Documents
Bohlender™
C425-83
6 mm 0.24 in. GL 25
245.00 NOK
Pack of 3
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
17187114
View Documents
Bohlender™
C425-86
10 mm 0.4 in. GL 25
241.00 NOK
Pack of 3
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
17107124
View Documents
Bohlender™
C425-90
16 mm 0.63 in. GL 25
133.00 NOK
Pack of 2
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
17197114
View Documents
Bohlender™
C425-88
16 mm 0.6 in. GL 32
201.00 NOK
Pack of 2
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
17117124
View Documents
Bohlender™
C425-92
22 mm 0.9 in. GL 32
179.00 NOK
Pack of 2
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
Description
Description
Screw caps compress the o-ring of the special gasket and provide sealing of the stirrer shaft.
Specifications
Specifications
Sealing Stirrer Shafts
PPS