Learn More
Bohlender™ BOLA™ Replacement Screw Caps, PPS
Replacement screw caps for sealing stirrer shafts.
Brand:  Bohlender™ C425-82
 View more versions of this product
Product Code. 17157114
  • 171.00 NOK / Pack of 3
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Description
Screw caps compress the o-ring of the special gasket and provide sealing of the stirrer shaft.
Specifications
Specifications
0.24 in.
Sealing Stirrer Shafts
GL 18
6 mm
PPS