Learn More
Bohlender™ BOLA™ Replacement Screw Caps, PPS
Replacement screw caps for sealing stirrer shafts.
Brand: Bohlender™ C425-83
This item is not returnable.
View return policy
Description
Screw caps compress the o-ring of the special gasket and provide sealing of the stirrer shaft.
Specifications
0.24 in. | |
Sealing Stirrer Shafts | |
GL 25 |
6 mm | |
PPS |