missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar ZINC, POWDER, 98% 2.5KG

Brand:  Thermo Scientific Alfa Aesar L13310.0E

ZINC, POWDER, 98% 2.5KG

Product Code. 11348026

  • 2220.00 NOK / 2500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252261 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberL13310.22,L13310.36,L13310.0E
Lot Number10248129 Certificate TypeCertificate of Analysis Date21/11/2023 Catalog NumberL13310.22,L13310.36,L13310.0E
Lot NumberY25G018 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL13310.22,L13310.36,L13310.0E
Lot NumberX13H029 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog NumberL13310.22,L13310.36,L13310.0E
Lot Number10242352 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog NumberL13310.22,L13310.36,L13310.0E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Alfa Aesar ZINC, POWDER, 98% 2.5KG >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.