Learn More
Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals
Description
Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to air and moisture. Incompatible with strong acids, strong bases and oxidizing agents.

Specifications
Specifications
Melting Point | 420°C |
Boiling Point | 907°C |
Density | 7.13 |
UN Number | UN1436 |
Quantity | 2.5 kg |
Solubility Information | Reacts with water. |
Percent Purity | ≥97% |
Mesh Size | -100 Mesh |
Odor | Odorless |
Sensitivity | Air and moisture sensitive |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y25G018 | Certificate of Analysis | 24/02/2023 | L13310.22,L13310.36,L13310.0E |
X13H029 | Certificate of Analysis | 19/09/2022 | L13310.22,L13310.36,L13310.0E |
10242352 | Certificate of Analysis | 25/07/2022 | L13310.22,L13310.36,L13310.0E |
10186560 | Certificate of Analysis | 21/05/2022 | L13310.22,L13310.36,L13310.0E |
10239952 | Certificate of Analysis | 12/04/2022 | L13310.22,L13310.36,L13310.0E |
Safety and Handling


- Zinc powder
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- MIXTURE LIST-Contains : Zinc powder
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.