missing translation for 'onlineSavingsMsg'
Learn More

YARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen YARS2
Concentration 0.27 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896984
View Documents Promotion Details
Proteintech
17150-1-AP-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16886984
View Documents Promotion Details
Proteintech
17150-1-AP-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
Specifications

Specifications

YARS2
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
Q5I0L3, Q8BYL4, Q9Y2Z4
287924, 51067, 70120
YARS2 Fusion Protein Ag9530
Primary
-20°C
YARS2
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CGI 04, mt TyrRS, Tyrosine tRNA ligase, TYRRS, YARS2
Yars2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications