missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17150-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).Specifications
YARS2 | |
Polyclonal | |
Unconjugated | |
YARS2 | |
CGI 04, mt TyrRS, Tyrosine tRNA ligase, TYRRS, YARS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287924, 51067, 70120 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5I0L3, Q8BYL4, Q9Y2Z4 | |
Yars2 | |
YARS2 Fusion Protein Ag9530 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |