Learn More
ABEBA™ X-Light Black Safety Trainers S1
ABEBA™ X-Light Black Safety Trainers S1 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
1165.00 NOK
Specifications
Certifications/Compliance | CE EN ISO 20345:2012 S2, SRC, ESD |
---|---|
Closure Type | Laces |
Color | Black |
Eyelets Material | Textile |
Liner Material | Breathable lining with Silver ions |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16358331
|
ABEBA™
7131038/35 |
35 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16368331
|
ABEBA™
7131038/36 |
36 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16378331
|
ABEBA™
7131038/37 |
37 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16388331
|
ABEBA™
7131038/38 |
38 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16398331
|
ABEBA™
7131038/39 |
39 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16308341
|
ABEBA™
7131038/40 |
40 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16318341
|
ABEBA™
7131038/41 |
41 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16328341
|
ABEBA™
7131038/42 |
42 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16338341
|
ABEBA™
7131038/43 |
43 |
1165.00 NOK
1 pair |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
16348341
|
ABEBA™
7131038/44 |
44 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16358341
|
ABEBA™
7131038/45 |
45 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16368341
|
ABEBA™
7131038/46 |
46 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16378341
|
ABEBA™
7131038/47 |
47 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
16388341
|
ABEBA™
7131038/48 |
48 |
1165.00 NOK
1 pair |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
Description
- Lightweight, comfort, hygiene
- Comply to EN ISO 20345 : 2012, S2 SRC
- ESD dissipative according to EN 61340 Climate class 1
- Slip resistant Mono PU outsole SRC
- Steel toe cap (200 joules)
- Black smooth leather
- Silver ions lining
- Replaceable Active Comfort insole
- Sanitized treatment
Specifications
CE EN ISO 20345:2012 S2, SRC, ESD | |
Black | |
Breathable lining with Silver ions | |
Slip and abrasion resistant, shock absorbing, ESD dissipative | |
Trainer | |
1 Pair | |
Water |
Laces | |
Textile | |
Leather | |
Polyurethane | |
Steel | |
CE EN ISO 20345:2012, DIN EN 61340 |
Product Title
ABEBA™ X-Light Black Safety Trainers S1