missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Wheaton™ Microlink Caps Product Code.: 12403881
Add to basket

DWK Life Sciences Wheaton™ Microlink Caps

Product Code. 12403881 Shop All Wheaton Science Products Products
38-430, Polypropylene/Silicone/PTFE, Black, 50 Cs.
Click to view available options
Neck Finish:
33-430
38-430
Closure Liner:
Polypropylene/Silicone
Polypropylene/Silicone/PTFE
Color:
Black
White
No. per Case:
24 Cs.
50 Cs.
This item is not returnable. View return policy

Product Code. 12403881

Brand: DWK Life Sciences 240114

Add to basket
Add to basket
This item is not returnable. View return policy

Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals

  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polypropylene
No. per Case 50 Cs.
Neck Finish 38-430
Certifications/Compliance ISO 9001
Closure Included Yes
Autoclavable Autoclavable
Product Type Microlink Cap
For Use With (Equipment) Bottle
Closure Liner Polypropylene/Silicone/PTFE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Microlink Caps >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.