Learn More
Fisherbrand™ SUB-GEL Midi-Plus UV Gel Scoop
Accessory for use with Horizontal SUB-GEL Midi-Plus Electrophoresis System.
Brand:  Fisherbrand™ MS15-UVS
Product Code. 11813423
  • 450.00 NOK / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for Horizontal SUB-GEL Midi-Plus Electrophoresis System
Order Info
dimForUseWithEquipment
Specifications
Specifications
150mm
Horizontal SUB-Gel Midi-Plus Electrophoresis System
SUB-GEL Midi-Plus UV Gel Scoop
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand and Fisher BioReagents Focus on electrophoresis
    Product Title
    Fisherbrand™ SUB-GEL Midi-Plus UV Gel Scoop >