missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LINGO-2 Antibody (382007), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36791J
This item is not returnable.
View return policy
Description
LINGO-2 Monoclonal antibody specifically detects LINGO-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
LINGO-2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
158038 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
382007 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LRRN6Cleucine rich repeat neuronal 6C | |
Mouse myeloma cell line NS0-derived recombinant human LINGO-2, Cys28-Leu542, Accession # Q7L985 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LINGO-2 Antibody (382007), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Spot an opportunity for improvement?Share a Content Correction