Learn More
Ismatec™ Tygon™ E-LFL L/S™ 2-Stop Precision Pump Tubing
Longest life of all Tygon™ formulations
Brand:  Ismatec™ 06447-16
 View more versions of this product
Product Code. 15309754
  • 2010.00 NOK / Pack of 8
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Long life - won't age or oxidize
  • Fits tightly to glass and metal & bends to small radius curves
  • Nontoxic
  • Transparent
  • For use with Masterflex™ multichannel pump heads
Specifications
Specifications
USP; RoHS; Non-Phthalate; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Prop 65 NA (no Prop 65 chemicals)
Tygon E-LFL
0.38 m
Pk. of 8
460 mL/min
40 psi, 2.7 bar
40°C
3.1 mm
1.25 ft.
0°C
16
1/8 in.
660 mm Hg
Stopped Pump Tubing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ismatec™ Tygon™ E-LFL L/S™ 2-Stop Precision Pump Tubing > 16