Learn More
Ismatec™ Tygon™ E-LFL L/S™ 2-Stop Precision Pump Tubing
Longest life of all Tygon™ formulations
Brand: Ismatec™ 06447-16
This item is not returnable.
View return policy
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Long life - won't age or oxidize
- Fits tightly to glass and metal & bends to small radius curves
- Nontoxic
- Transparent
- For use with Masterflex™ multichannel pump heads
Specifications
USP; RoHS; Non-Phthalate; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Prop 65 NA (no Prop 65 chemicals) | |
Tygon E-LFL | |
0.38 m | |
Pk. of 8 | |
460 mL/min | |
40 psi, 2.7 bar | |
40°C |
3.1 mm | |
1.25 ft. | |
0°C | |
16 | |
1/8 in. | |
660 mm Hg | |
Stopped Pump Tubing |
Product Content Correction
Product Title
Ismatec™ Tygon™ E-LFL L/S™ 2-Stop Precision Pump Tubing > 16