missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TSPO Recombinant Protein

4115.00 NOK - 6245.00 NOK

Specifications

For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molecular Weight (g/mol) 44.33
pH Range 8
Preparation Method In vitro wheat germ expression system
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16122401
View Documents Promotion Details
Abnova™
H00000706-P01.10ug
10 ug
4115.00 NOK
10µg
Please to purchase this item. Need a web account? Register with us today!
16132401
View Documents Promotion Details
Abnova™
H00000706-P01.25ug
25μg
6245.00 NOK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Antibody Production, Protein Array, ELISA, Western Blot
44.33
In vitro wheat germ expression system
Wheat Germ (in vitro)
Recombinant
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
8
Glutathione Sepharose 4 Fast Flow
Human
Solution
Videos
SDS
Documents

Documents

Product Certifications