missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TSPO Recombinant Protein
Brand: Abnova™ H00000706-P01.25ug
This item is not returnable.
View return policy
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
44.33 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
-80°C | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human TSPO Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human | |
Solution |