missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricine, ≥98.5% (Dry Basis), Ultrapure
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Brand: Thermo Scientific J22561.22
Quantity | 100 g |
---|---|
Packaging | Plastic bottle |
This item is not returnable.
View return policy
Description

Chemical Identifiers
5704-04-1 | |
179.17 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
79784 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
C6H13NO5 | |
MFCD00004277 | |
N-Tris(Hydroxymethyl)methylglycine | |
CHEBI:39063 | |
C(C(=O)O)NC(CO)(CO)CO |
Specifications
5704-04-1 | |
Crystalline Powder | |
C6H13NO5 | |
MFCD00004277 | |
1937804 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
79784 | |
179.17 | |
Dry basis |
∼183°C (decomposition) | |
100 g | |
(CH2OH)3CNHCH2COOH | |
Plastic bottle | |
N-Tris(Hydroxymethyl)methylglycine | |
C(C(=O)O)NC(CO)(CO)CO | |
179.17 | |
CHEBI:39063 | |
≥98.5% | |
Tricine |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
200946 | Certificate of Analysis | 13/11/2023 | J22561.36,J22561.22,J22561.A1 |
200946 | Certificate of Analysis | 27/11/2022 | J22561.36,J22561.22,J22561.A1 |
190764 | Certificate of Analysis | 27/11/2022 | J22561.36,J22561.22,J22561.A1 |
191502 | Certificate of Analysis | 27/11/2022 | J22561.36,J22561.22,J22561.A1 |
177505 | Certificate of Analysis | 27/11/2022 | J22561.36,J22561.22,J22561.A1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tricine, ≥98.5% (Dry Basis), Ultrapure > 100g; Plastic bottle
Spot an opportunity for improvement?Share a Content Correction