missing translation for 'onlineSavingsMsg'
Learn More

Tenovin-6, MedChemExpress

MedChemExpress Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).

Encompass_Preferred

1074.65 NOK - 8802.69 NOK

Specifications

Chemical Name or Material Tenovin-6
Physical Form Solid
CAS 1011557-82-6
Color Yellow
Purity Grade Notes Research
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16466786
Encompass_Preferred
View Documents
Medchem Express
HY-15510
100 mg
8802.69 NOK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16422677
Encompass_Preferred
View Documents
Medchem Express
HY-15510
50 mg
5215.24 NOK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16419157
Encompass_Preferred
View Documents
Medchem Express
HY-15510
5 mg
1438.14 NOK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16416588
Encompass_Preferred
View Documents
Medchem Express
HY-15510
10 mg
1801.63 NOK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16410246
Encompass_Preferred
View Documents
Medchem Express
HY-15510
2 mg
1074.65 NOK
2mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).
Specifications

Specifications

Tenovin-6
1011557-82-6
Research
Cancer-programmed cell death
O=C(NC(NC1=CC=C(NC(CCCCN(C)C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
454.63
Research
Solid
Yellow
C25H34N4O2S
DMSO : ≥ 31 mg/mL (68.19 mM)
454.63
98.67%
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Tenovin-6, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.