Learn More
Tenovin-6, MedChemExpress
MedChemExpress Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).
Brand: Medchem Express HY-15510
Description
Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).Specifications
Tenovin-6 | |
1011557-82-6 | |
10 mg | |
C25H34N4O2S | |
DMSO : ≥ 31 mg/mL (68.19 mM) | |
454.63 | |
98.67% |
Solid | |
Yellow | |
Research | |
Cancer-programmed cell death | |
O=C(NC(NC1=CC=C(NC(CCCCN(C)C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2 | |
454.63 | |
Research |