Learn More
Tedatioxetine hydrobromide, MedChemExpress
MedChemExpress Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist. ,

6858.82 NOK - 10936.19 NOK
Specifications
Chemical Name or Material | Tedatioxetine hydrobromide |
---|---|
CAS | 960151-65-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H22BrNS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16436277
![]() |
Medchem Express
HY-101755 |
5 mg |
6858.82 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16403558
![]() |
Medchem Express
HY-101755 |
10 mg |
10936.19 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist.Specifications
Tedatioxetine hydrobromide | |
White | |
C18H22BrNS | |
DMSO : 100 mg/mL (274.47 mM; Need ultrasonic) | |
364.34 | |
98.44% | |
Powder |
960151-65-9 | |
Research | |
Lu AA24530 hydrobromide | |
CC1=CC=C(SC2=CC=CC=C2C3CCNCC3)C=C1.[H]Br | |
364.34 | |
Research | |
Neuroscience-Neuromodulation |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.