Learn More
Tedatioxetine hydrobromide, MedChemExpress
MedChemExpress Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist. ,
Brand: Medchem Express HY-101755

Description
Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist.Specifications
Tedatioxetine hydrobromide | |
White | |
C18H22BrNS | |
Lu AA24530 hydrobromide | |
CC1=CC=C(SC2=CC=CC=C2C3CCNCC3)C=C1.[H]Br | |
364.34 | |
Research | |
Neuroscience-Neuromodulation |
960151-65-9 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (274.47 mM; Need ultrasonic) | |
364.34 | |
98.44% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only