missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC-E 5002
Selective KDM2/7 inhibitor
3190.00 NOK - 12460.00 NOK
Specifications
Purity | >98% |
---|---|
Product Type | TC-E 5002 |
Molecular Weight (g/mol) | 285.38 |
Inhibitors | TC-E 5002 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749508
|
Tocris Bioscience™
5089/10 |
10 mg |
3190.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787179
|
Tocris Bioscience™
5089/50 |
50 mg |
12460.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective histone demethylase KDM2/7 subfamily inhibitor (IC50 values are 0.2, 1.2, 6.8, 55, 83, >100 and >120 μM for KDM7A, KDM7B, KDM2A, KDM5A, KDM4C, KDM6A and KDM4A respectively). Inhibits growth of HeLa and KYSE-150 cancer cells in vitro.Specifications
>98% | |
TC-E 5002 | |
285.38 | |
TC-E 5002 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ TC-E 5002