Learn More
Tocris Bioscience™ TC-E 5002
Selective KDM2/7 inhibitor
Brand: Tocris Bioscience™ 5089/50
This item is not returnable.
View return policy
Description
TC-E 5002 is a selective histone demethylase KDM2/7 subfamily inhibitor (IC50 values are 0.2, 1.2, 6.8, 55, 83, >100 and >120 μM for KDM7A, KDM7B, KDM2A, KDM5A, KDM4C, KDM6A and KDM4A respectively). Inhibits growth of HeLa and KYSE-150 cancer cells in vitro.Specifications
>98% | |
TC-E 5002 | |
TC-E 5002 |
50 mg | |
285.38 |
Product Content Correction
Product Title
For Research Use Only