Learn More
Eppendorf™ Sterile Plate Lid
Brand: Eppendorf™ 0030131525
1983.90 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Specifications
Sterile | |
80 Each | |
Plate Lid |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
26Hours
7Minutes
16Seconds
52 -
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
Days
89Hours
7Minutes
16Seconds
52New Offer Save Up to 87% -
Multiple Manufacturers
Sample Preparation, Filtration and Storage
Get up to 80% off some of our most popular products.Promo Code: 24124Expires: 2025-08-31 Redemption Only
New Offer Save Now